Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ High Performance IEX Columns

Catalog No. 45002542 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
HiScreen Q Sepharose HP Column
HiScreen SP Sepharose HP Column
Ligand Type:
Quaternary Amine
Sulphopropyl
pH Range:
2 to 12
4 to 13
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type pH Range
45-002-542 HiScreen SP Sepharose HP Column Sulphopropyl 4 to 13
45-002-540 HiScreen Q Sepharose HP Column Quaternary Amine 2 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-542 Supplier Cytiva Supplier No. 28950515
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for method optimization and parameter screening

  • Ready-to-use columns for convenient process development optimization and parameter screening
  • Highly cross-linked agarose matrix with an average bead size of 34μm
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 6% cross-linked agarose beads
  • Average particle size: 34μm
  • Max. flow velocity: <150cm/hr.
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
  • Flow velocity: <150cm/hr.

HiScreen Q HP

  • Ionic capacity: 0.14 to 0.20mmol Cl-/mL
  • Ligand: quaternary amine
  • Ion exchanger type: strong anion exchanger
HiScreen SP HP
  • Ionic capacity: 0.15 to 0.20mmol Cl-/mL
  • Ligand: sulphopropyl
  • Ion exchanger type: strong cation exchanger

TRUSTED_SUSTAINABILITY

Specifications

Buffer 8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS
Product Type HiScreen SP Sepharose HP Column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <150 cm/hr.
Material Polypropylene
Ligand Type Sulphopropyl
Max. Pressure 3 bar
pH Range 4 to 13
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Optimization and parameter screening, ion exchange chromato graphy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.