Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Sepharose™ Prepacked Fast Flow HIC Columns

Catalog No. 45002538 Shop All Cytiva Products
Change view
Click to view available options
Stationary Phase:
Butyl Sepharose
Butyl-S-Sepharose
Octyl Sepharose
Phenyl (low sub) Sepharose
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
45-002-538 Phenyl (low sub) Sepharose
45-002-534 Butyl Sepharose
45-002-535 Butyl-S-Sepharose
45-002-536 Octyl Sepharose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-002-538 Supplier Cytiva Supplier No. 28926989
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for method optimization and parameter screening

  • Ready-to-use columns for convenient process development optimization
  • Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume of 4.7mL for fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with same chromatography media using same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 4% cross-linked agarose/6% cross-linked agarose
  • pH stability working range (media): 3 to 13
  • Average particle size: 90μm
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
  • Pressure max.: 1.5 bar (22psi)
  • Flow velocity: <240/450cm/hr.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 3M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6M Guanidine Hydrochloride, 8M Urea
Product Type HiScreen Phenyl (low sub) Sepharose 6 FF column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <450 cm/hr.
Material Polypropylene
Ligand Type Phenyl
Max. Pressure 1.5 bar
pH Range 3 to 13
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Stationary Phase Phenyl (low sub) Sepharose
Column Type Ion Exchange Column
For Use With (Application) Convenient process development optimization parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.