Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Sepharose™ Prepacked High Performance HIC Columns

Catalog No. 45002545 Shop All Cytiva Products
Change view
Click to view available options
Ligand Type:
Butyl
Phenyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ligand Type
45-002-545 Butyl
45-002-543 Phenyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-545 Supplier Cytiva Supplier No. 28978242
Add to Cart
Add to Cart

Designed for method optimization and parameter screening

  • Ready-to-use columns for convenient process development optimization
  • Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

HiScreen Phenyl HP

  • Degree of substitution: ∽25μmol phenyl/mL gel
HiScreen Butyl HP
  • Degree of substitution: ∽50μmol phenyl/mL gel

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 70% Ethanol, 3M Ammoniumsulfate, 1mM HCl, 30% Isopropanol, 2% SDS
Product Type HiScreen Butyl Sepharose HP column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7 mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <150 cm/hr.
Material Polypropylene
Ligand Type Butyl
Max. Pressure 3 bar
pH Range 3 to 13
Particle Size 24 to 44μm
Support Type Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Convenient process development optimization parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.