Learn More
CUBE BIOTECH Ready to use kit for His-tag protein purification kit, for 5x purifications
Supplier:  CUBE BIOTECH 80105
Encompass_Preferred
Ready to use kit for His-tag protein purification kit, for 5x purifications. 1x10 ml INDIGO Agarose, 2x50ml Binding Buffer, 2x50ml Washing Buffer, 1x50ml Elution buffer, 5 tubes, 1 x 500mM DTT, 1x500mM EDTA + Protocols
Catalog No. NC2684674
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.