Learn More
Invitrogen™ HIST2H2AB Polyclonal Antibody

Description
Predicted to react with rat, mouse, bovine and chicken based on sequence homology.
Specifications
Specifications
| Antigen | HIST2H2AB |
| Applications | ChIP Assay, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HIST2H2AB |
| Gene Accession No. | Q8IUE6 |
| Gene Alias | H2AB; HIST2H2AB; histone 2, H2ab; histone cluster 2, H2ab; histone H2A type 2-B |
| Gene Symbols | HIST2H2AB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.