Learn More
Selleck Chemical LLC Clofazimine S4107-1g
SDP
Supplier:  Selleck Chemical LLC S41071G
Encompass_Preferred
Clofazimine (NSC-141046) is a rhimophenazine dye originally developed for the treatment of tuberculosis it has both antimicrobial and antiinflammatory activity postulated mechanisms of action include intercalation of clofazimine with bacterial DNA and increasing levels of cellular phospholipase A2
Catalog No. 50-310-9388
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.