Learn More
Enzo Life Sciences Histamine polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2920100
Encompass_Preferred
Immunogen: Histamine conjugated to KLH. Source: Whole rabbit serum. Species reactivity: Species independent. Specificity: Recognizes histamine. Applications: ELISA, IHC. Formulation: Liquid. Neat rabbit serum.
Catalog No. 50-201-2832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.