Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Histamine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119564 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119564 Supplier Invitrogen™ Supplier No. PA5119564
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Histamine is located in mast cells, endocrine cells of the gut, blood cells and in some cells of the peripheral and central nervous system. Histamine is a potent vasodilator when secreted by mast cell found in various tissues as a result of allergic hytersensitivity or inflammation. In the central nervous system, Histamine is putative neurotransmitter. In the brain, its highest content has been found in the hypothalamus and in certain areas of the mesencephalon. The Histamine antiserum has a sensitivity level capable of detecting the low level Histamine contents of the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine
Applications Dot Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Host Species Rabbit
Immunogen Histamine.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.