Learn More
Novus Biologicals™ HistoGreen HRP Substrate
SDP
Catalog No. NB036828
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
NB036828 1 Kit
1 options
Catalog No. NB036828 Supplier Novus Biologicals™ Supplier No. E109
Customers require a valid business account associated to their profile to access pricing and ordering options.
very sensitive substrate chromogen for use in peroxidase-based immunohistochemical (IHC) and in situ-hybridisation (ISH) staining procedures
Specifications
Quantity 1 Kit
Substrate Type HistoGreen HRP Substrate
Storage Requirements Store at 4°C in the dark.