Learn More
VWR INTERNATIONAL LLC Histological Clearing Agent, 4 Liters
Supplier:  VWR INTERNATIONAL LLC 97060934
Encompass
Xylene replacement - non-carcinogenic and non-flammable Virtually odorless Faster wax dissolution and slower evaporation than xylene Detailed Description HistoChoice Clearing Agent is a much less hazardous replacement for xylene/toluene. Compatible with organic mounting media, HistoChoice Clearing Agent is designed to work with isopropanol in automated processors. It also offers faster wax dissolution and slower evaporation than xylene with no oily residue.
Catalog No. NC9948709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.