Learn More
Azer Scientific Histology Tissue Control Slides
Description
Includes
10 unstained and 1 stained Unifrost slide
- Prepared with paraffin-embedded human tissue sections
- For QC of laboratory's staining processes
Specifications
Specifications
| For Use With (Application) | Designed for use in monitoring stain performance in the pathology laboratory |
| Includes | 10 unstained and 1 stained unifrost slide |
| Quantity | 11 Pack |
| Product Type | ER Immunohistochemistry Control Slide |