Learn More
Johannes Lieder Histology of Vertebrata Microscope Slide Set
SCI_ED
Catalog No. S139709 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S139709 Cross and Longitudinal Section
1 options
This item is not returnable. View return policy
Catalog No. S139709 Supplier Johannes Lieder Supplier No. 2300
Add to Cart
Add to Cart
This item is not returnable. View return policy
25 microscope slides of the histology of Vertebrata, excluding Mammalia.
  • Fishes, Amphibians, Reptiles, and Birds.
  • 25 Microscope Slides
Specifications
Classification Histology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Slide Type Prepared Slide