Learn More
MBL International Corporation CycLex Cellular Histone Acetylation Assay Kit
SDP
Supplier:  MBL International Corporation CY1140
Encompass_Preferred
Size:2 x 96 Wells; Target:Cellular Histone Acetylation; The CycLex Cellular Histone Acetylation Assay Kit is used for the semi-quantitative measurement of histone acetylation level in situ by means of cell-based ELISA It can be used for 2 x 96 wells.CycLex Cellular Histone Acetylation Assay Kit.
Catalog No. 50-455-38
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.