Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Histone from Calf Thymus

Catalog No. ICN10072625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10072625 25 mg
ICN10072610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10072625 Supplier MP Biomedicals™ Supplier No. 0210072625
Add to Cart
Add to Cart

Arginine-rich subgroup

TRUSTED_SUSTAINABILITY

Specifications

Name Histone
Quantity 25 mg
Source Calf
Storage Requirements 2° to 8°C
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.