Learn More
Novus Biologicals™ Histone Deacetylase 4/HDAC4 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene ID (Entrez) | 9759 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56341 |
| Gene Alias | HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 |
| Gene Symbol | HDAC4 |
| Quantity | 0.1 mg |
| Research Category | Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.