Learn More
CPC Scientific H-Gly-Gly-Gly-Pro-Ala-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST011A
Encompass
This peptide is a substrate for CDK2 and CDK5 (cyclin dependent kinase 5) and PKA, can be phosphorylated by protein kinase A.ONE-LETTER SEQUENCE: GGGPATPKKAKKLMOLECULAR FORMULA: C56H101N17O15MOLECULAR WEIGHT:1252.51STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9441
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.