Learn More
CPC Scientific 5TAMRA-Gly-Gly-Gly-Pro-Ala-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST003B
Encompass
A TamrA-labeled histone H1-derived peptide (Ab/Em = 544/572 nm) that is phosphorylated by protein kinase A and serves as a substrate for CDK2 and CDK5 (cyclin dependent kinase 5) and PKA. ONE-LETTER SEQUENCE: 5TAMRA-GGGPATPKKAKKLMOLECULAR FORMULA: C81H121N19O19MOLECULAR WEIGHT:1665STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Pomerant, AM. et al. Proc. Natl. Acad. Sci. USA. 74, 4261 (1977)
Catalog No. 50-210-9427
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.