Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188794
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4095 10 μL
50-199-4101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4095 Supplier Active Motif Supplier No. 39708
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.

Specifications

Antigen Histone H1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene H1-2
Gene Accession No. P16401, P16403, Q02539
Gene Alias 0610008C09Rik; BOS_21872; BOS_21875; CTL-1; H1; H1 antibody; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.1; H1.2; H1.5; H10; H1-0; H1-1; H1a; H1b; H1c; H1f1; H1F2; H1F3; H1f5; H1FV; H1s-1; H1s-3; H1var1; H1var3; His1a; HIST1; HIST1H1A; Hist1h1b; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1.1; Histone H1.2; Histone H1.3; Histone H1.5; Histone H1a; Histone H1b; Histone H1c; Histone H1d; Histone H1s-1; Histone H1s-2; Histone H1s-3
Gene Symbols H1-1, H1-2, H1-5
Host Species Rabbit
Immunogen This Histone H1 antibody was raised against calf thymus histone H1.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3006, 3009, 3024
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.