Learn More
CPC Scientific H-Ala-Gln-Lys-Lys-Asp-Gly-Lys-Lys-Arg-Lys-Arg-Ser-Arg-Lys-Glu-Ser-Tyr-Ser-Ile-Tyr-Val-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST016B
Encompass
This peptide is the fragment of Histone 2B (21-41) with a C-terminal GG linker followed by a biotinylated lysine.ONE-LETTER SEQUENCE: AQKKDGKKRKRSRKESYSIYVGGK(Biotin)MOLECULAR FORMULA: C131H223N43O37S1MOLECULAR WEIGHT:3024.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9448
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.