Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B (phospho S14) Rabbit anti-Human, Clone: RM238, Abnova™

Catalog No. 89035322 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-322 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-322 Supplier Abnova Corporation Supplier No. MAB15114
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic phosphopeptide of human histone H2B (S14).

  • High specificity
  • High reactivity

Specifications

Antigen Histone H2B (phospho S14)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RM238
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic phosphopeptide of human histone H2B (S14).
Dilution ELISA (0.2-1 ug/mL) Immunocytochemistry (1-2 ug/mL) Multiplex (0.1-1 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P33778
Gene Alias H2B.1/H2B/f/H2BFF/MGC119804
Gene Symbols HIST1H2BB
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S14 of human histone H2B.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3018
Test Specificity This antibody reacts to histone H2B only when phosphorylated at S14. No cross reactivity with other phosphorylated histones.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.