Learn More
AnaSpec HISTONE H3 (1-21) BIOTINYLATE
Supplier:  AnaSpec AS61702
Encompass
Histone H3 (1-21), Biotinylated[ARTKQTARKSTGGKAPRKQLA-GG-K(BIOTIN)-NH2]; MW 2723.2; (1 mg) This sequence corresponds to the first 21 amino acids of the NH2 terminal of histone H3 followed by a GG linker and a biotinylated lysine. This peptide was used to investigate the characteristics and mechanisms of ethanol-induced histone H3 acetylation in rat hepatocytes. Immunocytochemical and immunoblot analyses revealed that ethanol treatment significantly increased H3 acetylation at Lys9 with negligible effects at Lys14, 18, and 23.
Catalog No. NC9786036
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.