Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Histone H3 Acetylation Assay Kit, Cell-Based, 48 Tests
Description
- Assay time: 5 hr
- Assay type: Cell-based
- Sensitivity: 1 ng
- Range: 1 ng to 100 ng
- Results type: Quantitative
- Applications: ELISA
- Sample types: Tissue, Suspension cells, Adherent cells
- Reactive species: Mouse, Human
- Instrument compatibility: Microplate reader
- Citations: 7
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Histone H3 (Acetylated) |
| No. of Reactions | 48 Tests |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Protein Stain Kit |
| Format | Kit |
| Detection Method | Colorimetric |
| Detection Location | In-Solution Detection |
| Cross Reactivity | Mouse, Human |
| Sample Type | Tissue, Suspension Cells, Adherent Cells |
| Quantity | 48 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?