Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 Acetylation Assay Kit, Cell-Based, Quantitative, 96 Tests

Catalog No. 502584861
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4861 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4861 Supplier Abcam Supplier No. AB11510296TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A quantitative cell-based assay kit for measuring Histone H3 acetylation in 5 hours.

  • Assay Type: Cell-based
  • Applications: ELISA
  • Sample Types: Tissue, Suspension cells, Adherent cells
  • Assay Time: 5 hr
  • Sensitivity: 1 ng
  • Detection Range: 1 ng to 100 ng
  • Results Type: Quantitative
  • Reactive Species: Mouse, Human
  • Instrument Compatibility: Microplate reader
  • Citations: 7

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Histone H3 (Acetylated)
Reactivity Human, Mouse
For Use With (Equipment) Microplate Reader
No. of Reactions 96 Tests
Product Type Western Blot Kit
Detection Location In-Solution Detection
Format Kit
Detection Method Colorimetric
Cross Reactivity Human, Mouse
Sample Type Tissue, Suspension Cells, Adherent Cells
Quantity 96 Tests
For Use With (Application) ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.