Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Histone H3 Blocking Peptide, Human, Synthetic, 1 mg
Description
- Target: Histone H3
- Type: Synthetic
- Applications: BL
- Species Reactivity: Human
- Form: Lyophilized
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Target Molecule | Histone H3 (tri methyl K9, phospho S10), Human |
| Reactivity | Human |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Label or Dye | Unconjugated |
| Quantity | 1 mg |
| For Use With (Application) | Blocking, Neutralization |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?