Learn More
Abcam Histone H3 (K4) Methyltransferase Activity Quantification Assay Kit, 48 Tests
Description
- Applications: Enzyme activity assay
- Sample types: Tissue Extracts, Cell Lysate
- Reactive species: Mouse, Human
- Assay time: 3 hr
- Results type: Quantitative
- Detection method: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Mouse, Human |
| Run Time | 3 hr. |
| Target Molecule | Histone H3 (K4) Methyltransferase |
| No. of Reactions | 48 Tests |
| For Use With (Equipment) | Microplate Reader |
| Detection Method | Colorimetric |
| Product Type | Western Blot Kit |
| Format | Kit |
| Cross Reactivity | Mouse, Human |
| Sample Type | Tissue Extracts, Cell Lysates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.