Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 (K4) Methyltransferase Activity Quantification Assay Kit, 96 Tests

Catalog No. 502584903
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4903 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4903 Supplier Abcam Supplier No. AB11345296TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantify Histone H3 (K4) methyltransferase activity in cell and tissue extracts in 3 hr.

  • Applications: Enzyme activity assay
  • Sample types: Tissue extracts and cell lysates
  • Assay time: 3 hr
  • Reactive species: Mouse, Human
  • Results type: Quantitative
  • Detected via: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Detection Method Colorimetric
Reactivity Human, Mouse
Target Molecule Histone H3 (K4) Methyltransferase, Human, Mouse
Run Time 3 hr.
Format Kit
Product Type Western Blot Kit
For Use With (Equipment) Microplate Reader
No. of Reactions 96 Tests
Cross Reactivity Mouse, Human
Sample Type Cell Lysates, Tissue Extracts
Quantity 96 Tests
For Use With (Application) Enzyme activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.