Learn More
Abcam Histone H3 (K9) Methyltransferase Activity Assay Kit, 48 Tests
Description
- Applications: Enzyme activity assay
- Sample Types: Tissue Extracts, Cell Lysate
- Species Reactivity: Mouse, Human
- Assay Time: 3 hr
- Results Type: Quantitative
- Key Features: Measures Histone H3 (K9) Methyltransferase activity; suitable for use with microplate reader; cited in > 2 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Histone H3 (K9) Methyltransferase |
| Run Time | 3 hr. |
| Reactivity | Human, Mouse |
| No. of Reactions | 48 Tests |
| Detection Location | In-Solution Detection |
| Product Type | Western Blot Kit |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Detection Method | Colorimetric |
| Cross Reactivity | Mouse, Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.