Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 K9 Methyltransferase Activity Assay Kit, 96 Tests

Catalog No. 502584901
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4901 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4901 Supplier Abcam Supplier No. AB11345396TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantifies histone H3 (K9) methyltransferase activity in cell or tissue extracts in 3 hr.

  • Applications: Enzyme activity assay
  • Sample types: Tissue extracts, Cell lysate
  • Reactive species: Mouse, Human
  • Assay time: 3 hr
  • Results type: Quantitative
  • Detection method: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Histone H3 (K9) Methyltransferase
For Use With (Equipment) Microplate Reader
No. of Reactions 96 Tests
Run Time 3 hr.
Format Kit
Detection Method Colorimetric
Detection Location In-Solution Detection
Cross Reactivity Mouse, Human
Sample Type Tissue Extracts, Cell Lysate
Quantity 96 Tests
For Use With (Application) Enzyme activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.