Learn More
Abcam Histone H3 Multiclonal Antibody, Rabbit Host, RM1001, 1 mL
Description
- Host species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Purification: Affinity purification Protein A
- Applications: IP, WB, ICC/IF, Dot, Flow Cyt (Intra), PepArr
- Species reactivity: Human, Mouse, Rat
- Form: Liquid
- Storage buffer: PBS, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide, pH 7.2 to 7.4
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Histone H3 (Citrullinated R2, R8, R17) |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Detection Location | In-Blot Detection |
| Buffer | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH7.2 to 7.4 |
| Detection Method | Chemiluminescence, Fluorescence |
| Cross Reactivity | Human, Mouse, Rat |
| Formulation | PBS, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide, pH 7.2 to 7.4 |
| Quantity | 1 mL |
| For Use With (Application) | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western blot, Dot blot, Peptide array |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.