Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 Peptide, Human, Di Methyl K9, 1 mg

Catalog No. 502704432
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4432 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4432 Supplier Abcam Supplier No. AB17721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Synthetic blocking peptide with >90% purity suitable for Western blot analysis.

  • Target Protein: Histone H3
  • Form: Liquid
  • Species Reactivity: Human
  • Purity: >90%
  • Tested Applications: BL
  • Composition: pH 7.4; 0.87% Sodium chloride, 0.476% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
  • Usage: Synthetic blocking peptide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Histone H3 (di methyl K9)
Reactivity Human
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Label or Dye Unconjugated
Form Liquid
Product Type Blocker and Diluent
Concentration 1 mg/mL
Content And Storage Store at -20°C.
Buffer 0.476% HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH7.4
Cross Reactivity Human
Formulation pH 7.4. Constituents: 0.87% Sodium chloride, 0.476% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated.
Quantity 1 mg
For Use With (Application) Blocking, Western blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.