Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 Peptide, Human, Tri-Methyl K27, Lyophilized, 500 ug

Catalog No. 502704429
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4429 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4429 Supplier Abcam Supplier No. AB1782500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Synthetic blocking peptide suitable for dot blot and western blot applications.

  • Purity: ≥70%
  • Form: Lyophilized
  • Tested Applications: BL, Dot
  • Species Reactivity: Human
  • Storage Conditions: Store at -20°C

Specifications

Target Molecule Histone H3 (Trimethyl K27)
Shipping Condition Blue Ice
Substrate Properties Peptide-based Substrate
Form Lyophilized
Reactivity Human
Product Type Blocker and Diluent
Label or Dye Unconjugated
Content And Storage Store at -20°C.
Detection Location In-Blot Detection
Cross Reactivity Human
Quantity 500 μg
For Use With (Application) Blocking, Dot Blot, Western Blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.