Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 Peptide, Synthetic, Human, Tri Methyl K4, 1 mg

Catalog No. 502707015
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7015 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7015 Supplier Abcam Supplier No. AB13421MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >90% purity, suitable for dot blot and Western blot applications.

Human Histone H3 (tri methyl K4) peptide is a synthetic blocking peptide with >90% purity.

  • Tested Applications: Dot, BL
  • Form: Lyophilized
  • Species Reactivity: Human

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Reactivity Human
Substrate Properties Peptide-based Substrate
Target Molecule Histone H3 (tri methyl K4)
Shipping Condition Blue Ice
Label or Dye Unconjugated
Form Lyophilized
Content And Storage Store at -20°C.
Detection Location In-Blot Detection
Detection Method X-ray Film or CCD Imager
Cross Reactivity Human
Quantity 1 mg
For Use With (Application) Blocking, Dot Blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.