Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 (phospho S10 + S28) Antibody, Rabbit Multiclonal, Carrier Free, 100 μg

Catalog No. 502691786
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-269-1786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-269-1786 Supplier Abcam Supplier No. AB303568100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit multiclonal antibody for the detection of histone H3 (phospho S10 + S28) in multiple applications including ChIP, flow cytometry, ICC/IF, IHC, and western blot.

  • Host Species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: PepArr, ChIP, Flow Cyt (Intra), WB, IHC-P, IHC-Fr, ICC/IF, Dot
  • Reactive Species: Human, Mouse, Rat
  • Purification: Affinity purification Protein A
  • Form: Liquid
  • Storage Buffer: pH 7.2 to 7.4, PBS

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Label or Dye Unconjugated
Target Molecule Histone H3 (Phosphoproteins (S10 + S28))
Buffer PBS, pH7.2 to pH7.4
Detection Location In-Blot Detection
Form Liquid
Detection Method Chemiluminescence, Fluorescence
Cross Reactivity Human, Mouse, Rat
Formulation PBS, pH 7.2 to 7.4
Quantity 100 μg
For Use With (Application) ChIP, Flow Cytometry, ICC/IF, IHC, Western Blot, Peptide Array, Dot Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.