Learn More
Abcam Histone H3 Total Acetylation Assay Kit, Fluorometric, Quantitative, 48 Tests
Description
- Assay Time: 2 hr 30 min
- Detection Method: Fluorescent
- Range: 0.4 ng to 50 ng
- Sensitivity: > 0.4 ng
- Results Type: Quantitative
- Sample Types: Tissue, Suspension cells, Adherent cells
- Reactive Species: Mouse, Human
- Compatible Equipment: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| No. of Reactions | 48 Tests |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Detection Location | In-Solution Detection |
| Product Type | Western Blot Kit |
| Target Molecule | Histone H3 (Acetylated) |
| Detection Method | Fluorescence |
| Cross Reactivity | Human, Mouse |
| Sample Type | Tissue, Suspension Cells, Adherent Cells |
| Quantity | 48 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.