Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H3 Total Acetylation Assay Kit, Fluorometric, Quantitative, 96 Tests

Catalog No. 502584067
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4067 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4067 Supplier Abcam Supplier No. AB13156196TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly detect and quantify total histone H3 acetylation in cell and tissue samples using a fluorometric assay.

  • Assay time: 2 hr 30 min
  • Detection method: Fluorescent
  • Range: 0.4 ng to 50 ng
  • Reactive species: Human, Mouse
  • Results type: Quantitative
  • Sample types: Adherent cells, Suspension cells, Tissue
  • Sensitivity: > 0.4 ng

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Protein-based Substrate
Target Molecule Histone H3 (Acetylated)
Run Time 2 hr. 30 min.
No. of Reactions 96 Tests
For Use With (Equipment) Microplate Reader
Product Type Protein Stain Kit
Detection Location In-Solution Detection
Detection Method Fluorescence
Format Kit
Cross Reactivity Mouse, Human
Sample Type Tissue, Suspension Cells, Adherent Cells
Quantity 96 Tests
For Use With (Application) Histone H3 Total Acetylation assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.