Learn More
Active Motif Histone H3 trimethyl Lys4 ELISA
SDP
Supplier:  Active Motif 53113
Encompass
Histone Modification ELISAs provide a simple, sensitive method for detecting changes in specific histone modifications from purified core histones or histones isolated by acid extraction. These kits are sandwich ELISAs that utilize a capture antibody against histone H3 and a primary antibody specifc for the modification of interest. A secondary antibody conjugated to horseradish peroxidase (HRP) and developing solutions provide a sensitive colorimetric readout that is easily quantified by spectrophotometry. The assay is performed in a convenient 96-stripwell plate, enabling low or high throughput screening.
Catalog No. NC1549009
Only null left
Edge
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.