Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3K4me3 Monoclonal Antibody (MABI 0304), Active Motif™
SDP

Catalog No. 501993357
Encompass
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3357 50 μg
50-199-3356 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3357 Supplier Active Motif Supplier No. 61979
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specifications

Antigen Histone H3K4me3
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Monoclonal
Clone MABI 0304
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene H3-3A
Gene Accession No. P68431, P84243, Q6NXT2, Q71DI3
Gene Alias B0035.7; BUR5; CELE_T10C6.12; CG31613; CG31613 gene product from transcript CG31613-RA; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K4; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3c1; H3c10; H3C11; H3C12; H3C2; H3C3; H3C4;
Gene Symbols H3-3A, H3-3B, H3-5, H3C1, H3C10, H3C14, H3C15, H3C2, H3C7
Host Species Mouse
Immunogen This Histone H3 trimethyl Lys4 antibody was raised against a synthetic peptide containing trimethyl Lys4 of human histone H3.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126961, 3020, 3021, 333932, 440093, 8350, 8357, 8358, 8968
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.