Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3S10ph Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188849
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3972 10 μL
50-199-3971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3972 Supplier Active Motif Supplier No. 39254
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specifications

Antigen Histone H3S10ph
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Host Species Rabbit
Immunogen This Histone H3 phospho Ser10 antibody was raised against a peptide including phospho-serine 10 of histone H3.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.