Learn More
Abcam Histone H4 Acetylation Assay Kit, Cell Based, Quantitative, 48 Tests
Description
- Assay Type: Cell-based
- Assay Time: 5 hr
- Sample Types: Tissue, Suspension cells, Adherent cells
- Reactive Species: Mouse, Human
- Results Type: Quantitative
- Detection Method: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Run Time | 5 hr. |
| Target Molecule | Histone H4 (Acetylated) |
| Reactivity | Human, Mouse |
| No. of Reactions | 48 Tests |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| Detection Method | Colorimetric |
| Cross Reactivity | Mouse, Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.