Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H4 Acetylation Assay Kit, Cell-based, Quantitative, 96 Test

Catalog No. 502708018
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-8018 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-8018 Supplier Abcam Supplier No. AB11510396TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid, quantitative cell-based assay kit for measuring histone H4 acetylation in tissue and cell samples within 5 hours.

  • Assay Type: Cell-based
  • Assay Time: 5 hr
  • Quantitative Results: Yes
  • Sample Types: Tissue, suspension cells, and adherent cells
  • Species Reactivity: Human and mouse
  • Detection Method: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Run Time 5 hr.
Target Molecule Histone H4 (Acetylated), Human, Mouse
For Use With (Equipment) Microplate Reader
No. of Reactions 96 Tests
Reactivity Human, Mouse
Format Kit
Product Type Western Blot Kit
Detection Method Colorimetric
Detection Location In-Solution Detection
Cross Reactivity Mouse, Human
Sample Type Tissue, Suspension Cells, Adherent Cells
Quantity 96 Tests
For Use With (Application) Histone H4 acetylation assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.