Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Histone H4 Peptide, Human, Synthetic, Unconjugated, 1 mg

Catalog No. 502707000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7000 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7000 Supplier Abcam Supplier No. AB138431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >90% purity, suitable for blocking applications.

  • Peptide Type: Synthetic
  • Species Reactivity: Human
  • Tested Applications: BL
  • Purity: >90%
  • Formulation: Liquid. Constituents: HEPES, 0.87% Sodium chloride, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
  • Storage: Store at -80°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Reactivity Human
Shipping Condition Blue Ice
Concentration 1 mg/mL
Form Liquid
Label or Dye Unconjugated
Target Molecule Histone H4 (Human)
Buffer HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated
Content And Storage Store at -80°C.
Cross Reactivity Human
Formulation HEPES, 0.87% Sodium chloride, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
Quantity 1 mg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.