Learn More
Histone H4H18ph Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H4H18ph |
| Applications | Dot Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against a synthetic peptide containing a 3-phosphohistidine analog at Histidine 18 of human Histone H4. |
| Purification Method | Protein A |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.