Learn More
Active Motif Histone H4K20me1 Peptide - biotinylated
SDP
Supplier:  Active Motif 81111
Encompass
Active Motif's biotinylated histone peptides are suitable for use in a variety of binding, pull-down and enzymatic assays for the study of proteins that interact with histones and that read, write or erase histone modifications. Histone H4K20me1 Peptide - biotinylated is a synthetic peptide derived from Histone H4, amino acids 1-23, with an added lysine residue containing a biotin moiety and a C-terminal amide group (CONH2). C-term biotinylated Lys residue is separated with two Gly (provide short spacer) and one Tyr (provides accurate UV quantification) from the rest of the peptide sequence. The presence of the tyrosine does not affect peptide functionality. Peptide Sequence: SGRGKGGKGLGKGGAKRHR-Kme-VLRGG-YK(Biotin)-NH2 Calculated MW Mavg 3005.56. Observed MW Mavg 3006.27. Purity: 95% by HPLC and Mass Spectrometry.
Catalog No. NC1851758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.