Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone macroH2A1.1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7189013
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2916 10 μg
50-199-2915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2916 Supplier Active Motif Supplier No. 39872
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

Specifications

Antigen Histone macroH2A1.1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene MACROH2A1
Gene Accession No. O75367
Gene Alias Core histone macro-H2A.1; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; H2AF12M; H2afy; Histone H2A.y; Histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; MACROH2A1; MACROH2A1.1; macroH2A1.2; Medulloblastoma antigen MU-MB-50.205; mH2A1
Gene Symbols MACROH2A1
Host Species Rabbit
Immunogen This macroH2A1.1 antibody was raised against a peptide corresponding to amino acids 198-226 of human macroH2A1.1.
Purification Method Antigen affinity chromatography
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9555
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.