Learn More
Cayman Chemical HIstonH2AZK7AcMoncLNal100ug
SDP
Supplier:  Cayman Chemical 32172100 UG
Encompass_Preferred
A major active metabolite of metamizole inhibits A23187-induced PGE2 production in isolated mouse peritoneal macrophages at 0.1 mM increases the paw withdrawal threshold in a rat model of carrageenan-induced hyperalgesia at 160 Ug/paw reduces LPS-induced pyrexia in rats at 60 90 and 120 mg/kg a potential impurity found in commercial preparations of metamizole
Catalog No. 50-294-9292
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.