Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoPrep™ Reagent Alcohol 70%
SDP

Catalog No. 11002218 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-218 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-218
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

70% reagent alcohol.

  • 100% reagent alcohol with 30 parts Type 1 water.
  • Used as a dehydrating agent during tissue processing, gradually removing water from the tissue sample before it is embedded in paraffin for sectioning and staining

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Grade Reagent

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.