Learn More
Abcam HistoReveal, Proteolytic Antigen Retrieval Reagent, Liquid, 15 mL
Description
- Proteolytic antigen retrieval (PIER) reagent
- Form: Liquid
- Storage buffer: 0.5% Sodium azide
- Storage temperature: +4°C
- Shipping temperature: Blue Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Form | Liquid |
| Alternate Part Number | ab103720 |
| Content And Storage | Store at 4°C. |
| Format | Bottle |
| Buffer | 0.5% sodium azide |
| Formulation | Preservative: 0.5% Sodium azide |
| Sample Type | Formalin-fixed paraffin-embedded sections, PFA-fixed paraffin-embedded sections |
| Quantity | 15 mL |
| For Use With (Application) | Proteolytic antigen retrieval |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.