Learn More
Epredia™ HistoScreen™ Cassettes
Catalog No. B851000BE Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851000BE Beige
B851000BL Blue
B851000GY Gray
B851000GN Green
B851000LA Lavender
B851000LG Light Green
B851000LI Lilac
B851000OR Orange
B851000PK Pink
B851000TN Tan
B851000WH White
B851000YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. B851000BE Supplier Epredia™ Supplier No. B851000BE
Add to Cart
Add to Cart
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
  • Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
  • Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
  • Precision engineering for solid connection to microtomes.
  • Cassette surface texture optimizes scanability for archiving.
Specifications
Pore Style Mesh
Closure Included Separate lids included
Quantity 1000/Cs.
Closure Type Back Hinge
Color Beige
Product Type HistoScreen™ Cassette
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov