Description
- Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
- Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
- Precision engineering for solid connection to microtomes.
- Cassette surface texture optimizes scanability for archiving.
Specifications
Specifications
| Pore Style | Mesh |
| Closure Included | Separate lids included |
| Quantity | 1000/Cs. |
| Closure Type | Back Hinge |
| Color | Pink |
| Product Type | HistoScreen™ Cassette |