Learn More
Simport™ Scientific Histosette™ I Tissue Processing/Embedding Cassette
Shop All Simport Scientific ProductsDescription
- Made of acetal
- Keeps specimens safely submerged in liquid
- Resistant to the chemical action of most histological solvents
- 30° angle anterior writing area
- One-piece construction with frangible hinges for easy lid removal
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
| Closure Included | Yes |
| Color | Lilac |
| Material | Acetal |
| Product Type | Tissue Cassette |
For in vitro diagnostic (IVD) use only.